ENAH3035CFE6 Enamine

tert-butyl N-{hexahydro-2'H-spiro[cyclobutane-1,1'-pyrrolo[1,2-a]piperazine]-3-yl}carbamate

  • MDL number MFCD31719362
  • Cas Number : 1934252-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-252295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-252295
Availability details for
Quantity
  • Availability
  • Add to Cart