ENAH3035D006 Enamine

ethyl 4-(2-nitrophenyl)-2,4-dioxobutanoate

  • MDL number MFCD07809017
  • Cas Number : 178114-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-258246

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-258246
Availability details for
Quantity
  • Availability
  • Add to Cart