ENAH3035D07A Enamine

[1,3]oxazolo[4,5-c]pyridin-2-amine

  • MDL number MFCD15526655
  • Cas Number : 114498-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-300657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-300657
Availability details for
Quantity
  • Availability
  • Add to Cart