ENAH3035D095 Enamine

2-amino-4-chloro-5-methylphenol

  • MDL number MFCD09038589
  • Cas Number : 53524-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-307015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-307015
Availability details for
Quantity
  • Availability
  • Add to Cart