ENAH3035D16D Enamine

(2E)-3-cyclobutylprop-2-enoic acid

  • MDL number MFCD06227778
  • Cas Number : 1355450-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-399888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-399888
Availability details for
Quantity
  • Availability
  • Add to Cart