ENAH3035D19F Enamine

1-methylpiperidine-3-carbonitrile

  • MDL number MFCD06658486
  • Cas Number : 4606-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-658418

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-658418
Availability details for
Quantity
  • Availability
  • Add to Cart