ENAH3035D490 Enamine

1-chloro-2-iodo-4-methoxybenzene

  • MDL number MFCD00070768
  • Cas Number : 2401-25-4
  • Molecular Weight : 268.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-2008855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008855
Availability details for
Quantity
  • Availability
  • Add to Cart