ENAH3035D935 Enamine

(2-iodoethoxy)trimethylsilane

  • MDL number MFCD20257314
  • Cas Number : 26305-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-4309916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4309916
Availability details for
Quantity
  • Availability
  • Add to Cart