ENAH3035D97B Enamine

4-bromo-1,2-thiazole-3-carbaldehyde

  • MDL number MFCD24676170
  • Cas Number : 88982-79-0
  • Molecular Weight : 192.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-4410755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4410755
Availability details for
Quantity
  • Availability
  • Add to Cart