ENAH3035DAD0 Enamine

3-formyl-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD22544381
  • Cas Number : 604001-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5469537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5469537
Availability details for
Quantity
  • Availability
  • Add to Cart