ENAH3035DDB1 Enamine

2-bromo-5-(bromomethyl)pyrazine

  • MDL number MFCD28134015
  • Cas Number : 1382866-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6245933

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-6245933
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart