ENAH3035DEA8 Enamine

4-chloro-2,1,3-benzothiadiazole

  • MDL number MFCD17168971
  • Cas Number : 2207-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6490061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6490061
Availability details for
Quantity
  • Availability
  • Add to Cart