ENAH3035E20D Enamine

3-(bromomethyl)-1,1-dimethoxycyclobutane

  • MDL number MFCD23106423
  • Cas Number : 1419101-20-4
  • Molecular Weight : 209.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6493223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6493223
Availability details for
Quantity
  • Availability
  • Add to Cart