ENAH3035E35D Enamine

methyl (1S,3R)-3-amino-2,2-dimethylcyclobutane-1-carboxylate hydrochloride

  • MDL number MFCD29925564
  • Cas Number : 2247088-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6494080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6494080
Availability details for
Quantity
  • Availability
  • Add to Cart