ENAH3035E54E Enamine

3-(tetramethyl-1,3,2-dioxaborolan-2-yl)furan-2-carbaldehyde

  • MDL number MFCD07368242
  • Cas Number : 1055881-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6495953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6495953
Availability details for
Quantity
  • Availability
  • Add to Cart