ENAH3035E708 Enamine

1-(3-aminophenyl)cyclopropane-1-carboxylic acid hydrobromide

  • MDL number MFCD31718773
  • Cas Number : 2248371-14-2
  • Molecular Weight : 258.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6497837

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6497837
Availability details for
Quantity
  • Availability
  • Add to Cart