ENAH3035E84C Enamine

1-bromo-5-chloro-4-iodo-2-methylbenzene

  • MDL number MFCD20233410
  • Cas Number : 1349718-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6495756

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6495756
Availability details for
Quantity
  • Availability
  • Add to Cart