ENAH3035EC5B Enamine

4-chloro-2H-chromen-2-one

  • MDL number MFCD00463528
  • Cas Number : 17831-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6501482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6501482
Availability details for
Quantity
  • Availability
  • Add to Cart