ENAH3035ED50 Enamine

decane-2,9-dione

  • MDL number MFCD03413036
  • Cas Number : 16538-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6504992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6504992
Availability details for
Quantity
  • Availability
  • Add to Cart