ENAH3035F379 Enamine

2,4-dimethylhexan-1-ol

  • MDL number MFCD21671571
  • Cas Number : 3965-59-1
  • Molecular Weight : 130.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6506322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6506322
Availability details for
Quantity
  • Availability
  • Add to Cart