ENAH3036065E Enamine

4-(difluoromethyl)thiophene-2-carboxylic acid

  • MDL number MFCD31915088
  • Cas Number : 2143554-23-6
  • Molecular Weight : 178.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6509361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6509361
Availability details for
Quantity
  • Availability
  • Add to Cart