ENAH3036816D Enamine

4-ethoxy-2-oxo-2H-pyran-6-carboxylic acid

  • MDL number MFCD00599926
  • Cas Number : 70158-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729029
Availability details for
Quantity
  • Availability
  • Add to Cart