ENAH3036816E Enamine

1-methanesulfinylpropan-2-ol

  • MDL number MFCD31719549
  • Cas Number : 21074-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729031

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729031
Availability details for
Quantity
  • Availability
  • Add to Cart