ENAH30368174 Enamine

tert-butyl N-[2-(methylsulfanyl)-1,3-thiazol-5-yl]carbamate

  • MDL number MFCD31719550
  • Cas Number : 2253631-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729060
Availability details for
Quantity
  • Availability
  • Add to Cart