ENAH30368176 Enamine

1-[2-(dimethylamino)ethyl]cyclopentane-1-carboxylic acid hydrochloride

  • MDL number MFCD31719551
  • Cas Number : 2253630-35-0
  • Molecular Weight : 221.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-(dimethylamino)ethyl)cyclopentane-1-carboxylic acid hydrochloride
Inchi Code 1S/C10H19NO2.ClH/c1-11(2)8-7-10(9(12)13)5-3-4-6-10;/h3-8H2,1-2H3,(H,12,13);1H
Inchi Key JWISRBZRPUNFSY-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6729063

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6729063
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart