ENAH30368177 Enamine

(2S)-2-[(2R)-2-amino-4-methylpentanamido]-3-methylbutanoic acid hydrochloride

  • MDL number MFCD31719552
  • Cas Number : 2253619-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729065
Availability details for
Quantity
  • Availability
  • Add to Cart