ENAH3036817C Enamine

2-(3-bromophenyl)-2-cyclohexylacetic acid

  • MDL number MFCD16457234
  • Cas Number : 1225786-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729193
Availability details for
Quantity
  • Availability
  • Add to Cart