ENAH3036817D Enamine

7-oxa-3lambda6-thia-10-azaspiro[5.6]dodecane-3,3-dione hydrochloride

  • MDL number MFCD31719555
  • Cas Number : 2253640-57-0
  • Molecular Weight : 255.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-oxa-3-thia-10-azaspiro[5.6]dodecane 3,3-dioxide hydrochloride
Inchi Code 1S/C9H17NO3S.ClH/c11-14(12)7-2-9(3-8-14)1-4-10-5-6-13-9;/h10H,1-8H2;1H
Inchi Key SKIRZDUKRCISAP-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6729201

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6729201
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart