ENAH30368181 Enamine

7-oxa-3-thia-10-azaspiro[5.6]dodecane hydrochloride

  • MDL number MFCD31719557
  • Cas Number : 2253641-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729206
Availability details for
Quantity
  • Availability
  • Add to Cart