ENAH30368185 Enamine

5-(chloromethyl)-2,5'-bipyrimidine hydrochloride

  • MDL number MFCD31719559
  • Cas Number : 2253631-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729228
Availability details for
Quantity
  • Availability
  • Add to Cart