ENAH30368189 Enamine

4-[(5-methyl-1,3,4-oxadiazol-2-yl)methyl]benzoic acid

  • MDL number MFCD24498201
  • Cas Number : 936755-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729358
Availability details for
Quantity
  • Availability
  • Add to Cart