ENAH3036818B Enamine

5-(ethoxycarbonyl)-3,4-dimethylthiophene-2-carboxylic acid

  • MDL number MFCD30294026
  • Cas Number : 94785-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231-233
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729360
Availability details for
Quantity
  • Availability
  • Add to Cart