ENAH30368192 Enamine

2-(4-fluoro-2-nitrophenyl)acetonitrile

  • MDL number MFCD13194395
  • Cas Number : 708-58-7
  • Molecular Weight : 180.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 46-50
Iupac Name 2-(4-fluoro-2-nitrophenyl)acetonitrile
Inchi Code 1S/C8H5FN2O2/c9-7-2-1-6(3-4-10)8(5-7)11(12)13/h1-2,5H,3H2
Inchi Key QRMIXFQTVUPDSM-UHFFFAOYSA-N

Description

Melting Point 46-50
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6729379

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6729379
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart