ENAH30368194 Enamine

potassium 1-hydroxypiperidine-2-carboxylate

  • MDL number MFCD31719582
  • Cas Number : 2253632-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729421
Availability details for
Quantity
  • Availability
  • Add to Cart