ENAH30368195 Enamine

5-formyl-1,2,3,4-tetrahydropyridine-2-carboxylic acid

  • MDL number MFCD31719583
  • Cas Number : 2253639-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729438
Availability details for
Quantity
  • Availability
  • Add to Cart