ENAH30368196 Enamine

(4-tert-butyl-1,3-thiazol-2-yl)(4-chlorophenyl)methanamine

  • MDL number MFCD13496196
  • Cas Number : 1305779-05-8
  • Molecular Weight : 280.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-(tert-butyl)thiazol-2-yl)(4-chlorophenyl)methanamine
Inchi Code 1S/C14H17ClN2S/c1-14(2,3)11-8-18-13(17-11)12(16)9-4-6-10(15)7-5-9/h4-8,12H,16H2,1-3H3
Inchi Key SVELVVDYXZGAEJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6729448

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6729448
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart