ENAH303681A0 Enamine

4-(4-chlorophenyl)-4-(dimethylamino)cyclohexan-1-one

  • MDL number MFCD28501826
  • Cas Number : 65618-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-111
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729509
Availability details for
Quantity
  • Availability
  • Add to Cart