ENAH303681AB Enamine

7-(aminomethyl)-1,2-dihydro-1,8-naphthyridin-2-one hydrochloride

  • MDL number MFCD31719366
  • Cas Number : 2253640-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729575
Availability details for
Quantity
  • Availability
  • Add to Cart