ENAH303681AD Enamine

methyl (1r,3r)-3-(5-formyl-1H-pyrazol-1-yl)cyclobutane-1-carboxylate

  • MDL number MFCD31719367
  • Cas Number : 2253619-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729595
Availability details for
Quantity
  • Availability
  • Add to Cart