ENAH303681B5 Enamine

3-bromo-2-chloro-5-(difluoromethoxy)pyridine

  • MDL number MFCD28733845
  • Cas Number : 130115-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729672
Availability details for
Quantity
  • Availability
  • Add to Cart