ENAH303681B8 Enamine

1-(5-methoxypyridin-3-yl)ethan-1-one hydrochloride

  • MDL number MFCD31719568
  • Cas Number : 2253630-37-2
  • Molecular Weight : 187.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(5-methoxypyridin-3-yl)ethan-1-one hydrochloride
Inchi Code 1S/C8H9NO2.ClH/c1-6(10)7-3-8(11-2)5-9-4-7;/h3-5H,1-2H3;1H
Inchi Key ZREAIQNQRWJOFG-UHFFFAOYSA-N

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6729708

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6729708
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart