ENAH303681BF Enamine

{6-aminospiro[3.3]heptan-2-yl}methanol hydrochloride

  • MDL number MFCD20718100
  • Cas Number : 170508-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729731
Availability details for
Quantity
  • Availability
  • Add to Cart