ENAH303681C4 Enamine

3-amino-2-(methoxymethyl)propane-1,2-diol hydrochloride

  • MDL number MFCD31719375
  • Cas Number : 2253639-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729744
Availability details for
Quantity
  • Availability
  • Add to Cart