ENAH303681C9 Enamine

tert-butyl 5,6,7,8-tetrahydropteridine-8-carboxylate

  • MDL number MFCD29048093
  • Cas Number : 1936361-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729781
Availability details for
Quantity
  • Availability
  • Add to Cart