ENAH303681D1 Enamine

benzyl 2-{[(tert-butoxy)carbonyl](methyl)amino}prop-2-enoate

  • Molecular Weight : 291.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 2-((tert-butoxycarbonyl)(methyl)amino)acrylate
Inchi Code 1S/C16H21NO4/c1-12(17(5)15(19)21-16(2,3)4)14(18)20-11-13-9-7-6-8-10-13/h6-10H,1,11H2,2-5H3
Inchi Key PPQBFRMCUVRASG-UHFFFAOYSA-N

Description

Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6729830
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart