ENAH303681DA Enamine

6-bromo-5-chloro-2-methylpyridin-3-amine

  • MDL number MFCD31655734
  • Cas Number : 2115000-36-5
  • Molecular Weight : 221.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-5-chloro-2-methylpyridin-3-amine
Inchi Code 1S/C6H6BrClN2/c1-3-5(9)2-4(8)6(7)10-3/h2H,9H2,1H3
Inchi Key GIAPXABRAYBXJA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6729872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729872
Availability details for
Quantity
  • Availability
  • Add to Cart