ENAH303681DB Enamine

5,6-dichloro-2-methylpyridin-3-amine

  • MDL number MFCD31719615
  • Cas Number : 2230279-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729875
Availability details for
Quantity
  • Availability
  • Add to Cart