ENAH303681DC Enamine

2-(2,6-dimethoxybenzoyl)-5-methylfuran

  • MDL number MFCD16513609
  • Cas Number : 1281546-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729876
Availability details for
Quantity
  • Availability
  • Add to Cart