ENAH303681DF Enamine

ethyl 2-[3-(difluoromethyl)phenyl]acetate

  • MDL number MFCD31719574
  • Cas Number : 2164055-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729885
Availability details for
Quantity
  • Availability
  • Add to Cart