ENAH303681E0 Enamine

2-{[2-(pyrrolidin-1-yl)ethyl]amino}propanoic acid dihydrochloride

  • MDL number MFCD31719616

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6729886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6729886
Availability details for
Quantity
  • Availability
  • Add to Cart